n-Dodecyl β-D-maltoside
Synonym(s):
DDM, Lauryl-β-D-maltoside, n-Dodecyl-β-D-maltoside, ULTROL® Grade
Empirical Formula (Hill Notation):
C24H46O11
CAS Number:
Molecular Weight:
510.62
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing