Skip to Content
MilliporeSigma
(3RS)-9-Fluoro-3-methyl-10-(4-methyl-4-oxidopiperazin-1-yl)-7-oxo-2,3-dihydro-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

(3RS)-9-Fluoro-3-methyl-10-(4-methyl-4-oxidopiperazin-1-yl)-7-oxo-2,3-dihydro-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid

(3RS)-9-Fluoro-3-methyl-10-(4-methyl-4-oxidopiperazin-1-yl)-7-oxo-2,3-dihydro-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Empirical Formula (Hill Notation):
C18H20FN3O5
CAS Number:
Molecular Weight:
377.37

Products

Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade