Skip to Content
MilliporeSigma
Prostaglandin E1 ≥98% (HPLC), synthetic

Prostaglandin E1

Prostaglandin E1 ≥98% (HPLC), synthetic
Synonym(s):
(11α,13E,15S)-11,15-Dihydroxy-9-oxoprost-13-enoic acid, Alprostadil, PGE1, Prostaglandin E1
Empirical Formula (Hill Notation):
C20H34O5
CAS Number:
Molecular Weight:
354.48

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Product Line
Purity
Research Area
Quality Segment
Sterilization Method
Technique