Skip to Content
MilliporeSigma
2-[5-(1H-1,2,4-Triazol-1-ylmethyl)-1H-indol-3-yl]ethanol certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

2-[5-(1H-1,2,4-Triazol-1-ylmethyl)-1H-indol-3-yl]ethanol

2-[5-(1H-1,2,4-Triazol-1-ylmethyl)-1H-indol-3-yl]ethanol certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Empirical Formula (Hill Notation):
C13H14N4O
CAS Number:
Molecular Weight:
242.28

Products

Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Quality Segment
Application
API Family
Reference Material Type
Special Grade