Skip to Content
MilliporeSigma
Melatonin powder, ≥98% (TLC)

Melatonin

Melatonin powder, ≥98% (TLC)
Synonym(s):
N-Acetyl-5-methoxytryptamine, Melatonine, Melatonin
Empirical Formula (Hill Notation):
C13H16N2O2
CAS Number:
Molecular Weight:
232.28

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Research Area
Quality Segment