Skip to Content
MilliporeSigma
2-[2-[[3-[2-(Dimethylamino)ethyl]-1H-indol-5-yl]methyl]-5-(1H-1,2,4-triazol-1-ylmethyl)-1H-indol-3-yl]-N,N-dimethylethanamine certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

2-[2-[[3-[2-(Dimethylamino)ethyl]-1H-indol-5-yl]methyl]-5-(1H-1,2,4-triazol-1-ylmethyl)-1H-indol-3-yl]-N,N-dimethylethanamine

2-[2-[[3-[2-(Dimethylamino)ethyl]-1H-indol-5-yl]methyl]-5-(1H-1,2,4-triazol-1-ylmethyl)-1H-indol-3-yl]-N,N-dimethylethanamine certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Empirical Formula (Hill Notation):
C28H35N7
CAS Number:
Molecular Weight:
469.62

Products

Brand
Formula Weight
Manufacturer
Product Line
Application
Special Grade