Skip to Content
MilliporeSigma
3-Methyl-1-phenyl-2-pyrazoline-5-one 99%

3-Methyl-1-phenyl-2-pyrazoline-5-one

3-Methyl-1-phenyl-2-pyrazoline-5-one 99%
Synonym(s):
5-Methyl-2-phenyl-2,4-dihydro-3H-pyrazol-3-one, MCI-186
Empirical Formula (Hill Notation):
C10H10N2O
CAS Number:
Molecular Weight:
174.20

Products

Product Category
Brand
Boiling Point (°C)
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Research Area
Quality Segment
Application
Markush Class
Markush Group