Skip to Content
MilliporeSigma
Salicylic acid meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)

Salicylic acid

Salicylic acid meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Synonym(s):
2-Hydroxybenzoic acid, Salicylic acid
Linear Formula:
2-(HO)C6H4CO2H
CAS Number:
Molecular Weight:
138.12
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Technique