L-Isoleucine
Synonym(s):
(2S,3S)-2-Amino-3-methylpentanoic acid, L-Isoleucine, (S)-(+)-Isoleucine
Linear Formula:
C2H5CH(CH3)CH(NH2)CO2H
CAS Number:
Molecular Weight:
131.17
Products
Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Optical Isomer
Amino Acid
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing