Skip to Content
MilliporeSigma
Hydroxypropylbetadex European Pharmacopoeia (EP) Reference Standard

(2-Hydroxypropyl)-β-cyclodextrin

Hydroxypropylbetadex European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Cyclodextrin HPB, (2-Hydroxypropyl)-β-cyclodextrin
Linear Formula:
C42H70-nO35 · (C3H7O)n
CAS Number:

Products

Product Category
Brand
Biological Source
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade