Skip to Content
MilliporeSigma
Chloroform-d "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer

Chloroform-d

Chloroform-d "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Synonym(s):
Deuterochloroform, Chloroform-D1
Linear Formula:
CDCl3
CAS Number:
Molecular Weight:
120.38
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Isotope
Technique