MilliporeSigma
Carbohydrates
Carbohydrates play many important roles in living organisms. They are a key nutrients, serving as the main source of energy and are involved in various biochemical reactions related to diseases and immune responses.
Carbohydrates, also called saccharides, can be divided into four groups: monosaccharides, disaccharides, oligosaccharides and polysaccharides. Monosaccharides are made of one unit of a saccharide; one example is glucose. Disaccharides are made of two monosaccharides linked together, such as lactose. Oligosaccharides are made of 3-9 monosaccharides and polysaccharides are composed of more than 9 monosaccharides linked together, such as starch.
Glycans are monosaccharides linked together by glycosidic bonds in chain-like structures. They are present on the surface of cells and are involved in many biochemical pathways from helping cells to communicate to recognizing invading pathogens. They are diverse, complex molecules and their functions are determined by their unique structures.
We offer a broad portfolio of highly pure carbohydrates for metabolomics, glycobiology/glycomics, microbiome, and nutrition research. Our carbohydrates are key supplements in cell culture media and are widely used as building blocks in the synthesis of glycoconjugates and glycans. Whatever your application, you can rely on our high-quality products for reliable, reproducible results.