Skip to Content
MilliporeSigma
All Photos(3)

Documents

V900336

Sigma-Aldrich

MES monohydrate

Vetec, reagent grade, 99%

Synonym(s):

2-(N-Morpholino)ethanesulfonic acid, 4-Morpholineethanesulfonic acid monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H13NO4S · H2O
CAS Number:
Molecular Weight:
213.25
Beilstein:
6350956
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

99%

form

powder or crystals

pH

2.5-4 (25 °C, 97.6 g/L)

useful pH range

5.5-6.7

pKa (25 °C)

6.1

mp

>300 °C (lit.)

solubility

water: 1.066 g/10 mL, clear, colorless

SMILES string

O.OS(=O)(=O)CCN1CCOCC1

InChI

1S/C6H13NO4S.H2O/c8-12(9,10)6-3-7-1-4-11-5-2-7;/h1-6H2,(H,8,9,10);1H2

InChI key

MIIIXQJBDGSIKL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

MES monohydrate (MES) is one of the "Good′s" buffers developed for biological applications.

Application

MES monohydrate (MES) can be used:
  • To maintain the pH during the hyaluronic acid functionalization using catechol.
  • In the synthesis of CaO2/MnO2 composites by the reduction of KMnO4.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

pH responsive chitosan and hyaluronic acid layer by layer film for drug delivery applications
Lu B, et al.
Progress in Organic Coatings, 135(2), 240-247 (2019)
Self-production of oxygen system CaO2/MnO2@ PDA-MB for the photodynamic therapy research and switch-control tumor cell imaging
Ji C, et al.
Journal of Biomedical Materials Research. Part B, Applied Biomaterials, 106(7), 2544-2552 (2018)
Hydrogen ion buffers for biological research.
Good NE, et al.
Biochemistry, 5(2), 467-477 (1966)
Tongtong Ma et al.
International journal of pharmaceutics, 473(1-2), 296-303 (2014-07-02)
Oral delivery of antigens is patient-friendly and efficient way of treating intestinal infections. However, the efficacy of oral vaccines is limited by degradation in the gastrointestinal (GI) tract and poor absorption by enterocytes and antigen-presenting cells (APC). Here we report
Tomoya Narawa et al.
Biological & pharmaceutical bulletin, 38(4), 545-551 (2015-04-04)
The stereoselective transport of methotrexate (L-amethopterin, L-MTX) and its enantiomer (D-amethopterin, D-MTX) by the rat proton-coupled folate transporter (rPCFT) were examined using rPCFT-expressing HEK293 cells. The initial rate of uptake of [3H]-L-MTX by the rPCFT followed Michaelis-Menten kinetics, with a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service