Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP0114

Sigma-Aldrich

NCOR2/SMRT human

recombinant, expressed in E. coli, ≥60% (SDS-PAGE)

Synonym(s):

Nuclear receptor corepressor 2, SMRTe, Silencing mediator of retinoic acid and thyroid hormone receptor, T3 receptor-associating factor, TRAC-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥60% (SDS-PAGE)

form

aqueous solution

mol wt

39 kDa

packaging

pkg of 50 μg

concentration

>0.02 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... NCOR2(9612)

General description

Human NCOR2/SMRT, DAD domain, amino acid 395-489, GenBank Accession No. NM_006312, N-terminal ST tag, MW = 39 kDa, expressed in E. coli system. NcoR2, nuclear receptor corepressor, are transcriptional corepressors.

Application

Useful for the study of enzyme kinetics, screening inhibitors and selectivity profiling.

Physical form

Formulated in 50 mM Tris-HCl, pH 8.0, 138 mM NaCl, 20 mM glutathione, and 10% glycerol.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service