Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4500535

Sigma-Aldrich

Anti-CTBP2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

C-terminal binding protein 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 48 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CTBP2(1488)

General description

The gene CTBP2 (C-terminal binding protein 2) encodes a member of the CtBP-family. The gene is mapped to human chromosome 21q21.3. It is found to be expressed ubiquitously, with higher expression in the heart, skeletal muscle, and pancreas.

Immunogen

The antiserum was produced against synthesized peptide derived from human CtBP2.

Immunogen Range: 396-445

Biochem/physiol Actions

The gene CTBP2 (C-terminal binding protein 2) encodes a protein that functions as a transcriptional co-repressor of several tumor suppressor genes resulting in enhanced cancer cell migration and invasion. Its expression is found to be upregulated in hepatocellular carcinoma (HCC). It may be a potential prognostic marker for post liver resection HCC. It is involved in several types of tumor initiation, progression and response to therapy. It is found to interact with the C-terminal region of adenovirus type 2/5 E1A protein, a region that negatively regulates tumorigenicity and the extent of oncogenic transformation.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel C-terminal binding protein (CTBP2) is closely related to CTBP1, an adenovirus E1A-binding protein, and maps to human chromosome 21q21.3.
Katsanis N and Fisher EM
Genomics, 47, 294-299 (1998)
Xin Zheng et al.
Oncotarget, 6(6), 3752-3769 (2015-02-18)
C-terminal binding protein 2 (CtBP2) is a transcriptional co-repressor that promotes cancer cell migration and invasion by inhibiting multiple tumor suppressor genes that contribute to cell mobility and adhesion. In this investigation, we showed thatCtBP2 expression was increased significantly in
CtBP2 could promote prostate cancer cell proliferation through c-Myc signaling.
Zhang C
Gene, 546, 73-79 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service