Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB3500952

Sigma-Aldrich

Anti-IL-37 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 24 kDa

species reactivity

human

concentration

1 mg/mL

technique(s)

ELISA: suitable
immunoblotting: suitable
immunofluorescence: suitable
immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL37(27178)

General description

IL-37 (interleukin 37) is an anti-inflammatory cytokine and is also termed as IL-1 family member 7 (IL-1F7). IL-37 belongs to the IL-1 cytokine family. This protein is expressed in blood monocytes, endothelial cells, adipocytes and epithelial cells. IL-37 is located on human chromosome 2q12.

Immunogen

Antibody was raised against a 17 amino acid peptide near the amino terminus of human IL-37.

Biochem/physiol Actions

IL-37 (interleukin 37) acts as a suppressor for innate immunity and acquired immunity. IL-37 participates in the immunosuppressive property of human CD4+CD25+ regulatory T cells. It represses the synthesis of several pro-inflammatory cytokines like IL-1α, IL-1β, IL-6, IL-12, granulocyte colony-stimulating factor (G-SCF), granulocyte-macrophage colony-stimulating factor (GM-CSF) and TNF-α (tumor necrosis factor alpha). It plays an important role in adaptive immunity.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500952.

Physical form

Supplied at 1 mg/mL in PBS with 0.02% sodim azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The interleukin-1 family: back to the future.
Garlanda C, et al.
Immunity, 39(6), 1003-1018 (2013)
Diana Boraschi et al.
European cytokine network, 22(3), 127-147 (2011-11-04)
The IL-1 family of cytokines encompasses eleven proteins that each share a similar β-barrel structure and bind to Ig-like receptors. Some of the IL-1-like cytokines have been well characterised, and play key roles in the development and regulation of inflammation.
Association between interleukin 37 (rs3811047) polymorphism and multiple autoimmune diseases in a Chinese population: A PRISMA-compliant meta-analysis.
Lin XY, et al.
Medicine, 97(15), e0386-e0386 (2018)
Interleukin-1 as a Key Factor in the Development of Inflammatory Diseases.
Farivar S, et al.
Archives of Pediatric Infectious Diseases, 2(4), e18177-e18177 (2014)
Macrophage-Specific Expression of IL-37 in Hyperlipidemic Mice Attenuates Atherosclerosis.
McCurdy S, et al.
Journal of Immunology, 199(10), 3604-3613 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service