Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1411672

Sigma-Aldrich

Anti-TP53I3 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

PIG3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 36.63 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TP53I3(9540)

General description

The protein encoded by this gene is similar to oxidoreductases, which are enzymes involved in cellular responses to oxidative stresses and irradiation. This gene is induced by the tumor suppressor p53 and is thought to be involved in p53-mediated cell death. It contains a p53 consensus binding site in its promoter region and a downstream pentanucleotide microsatellite sequence. P53 has been shown to transcriptionally activate this gene by interacting with the downstream pentanucleotide microsatellite sequence. The microsatellite is polymorphic, with a varying number of pentanucleotide repeats directly correlated with the extent of transcriptional activation by p53. It has been suggested that the microsatellite polymorphism may be associated with differential susceptibility to cancer. At least two transcript variants encoding the same protein have been found for this gene. (provided by RefSeq)

Immunogen

TP53I3 (AAH00474.1, 1 a.a. ~ 332 a.a) full-length human protein.

Sequence
MLAVHFDKPGGPENLYVKEVAKPSPGEGEVLLKVAASALNRADLMQRQGQYDPPPGASNILGLEASGHVAELGPGCQGHWKIGDTAMALLPGGGQAQYVTVPEGLLMPIPEGLTLTQAAAIPEAWLTAFQLLHLVGNVQAGDYVLIHAGLSGVGTAAIQLTRMAGAIPLVTAGSQKKLQMAEKLGAAAGFNYKKEDFSEATLKFTKGAGVNLILDCIGGSYWEKNVNCLALDGRWVLYGLMGGGDINGPLFSKLLFKRGSLITSLLRSRDNKYKQMLVNAFTEQILPHFSTEGPQRLLPVLDRIYPVTEIQEAHKYMEANKNIGKIVLELPQ

Biochem/physiol Actions

TP53I3 (Tumor protein p53 inducible protein 3) plays an important role in the generation of reactive oxygen species (ROS) and the programmed cell death. It functions as a pro-apoptotic marker in the apoptotic pathway. It has been suggested that PIG3 (p53-inducible gene 3) may initiate oxidative stress, like other p53 targets. It mediates apoptosis by generating reactive oxygen species through oxidative stress.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sergio Porté et al.
The Journal of biological chemistry, 284(25), 17194-17205 (2009-04-08)
Tumor suppressor p53 regulates the expression of p53-induced genes (PIG) that trigger apoptosis. PIG3 or TP53I3 is the only known member of the medium chain dehydrogenase/reductase superfamily induced by p53 and is used as a proapoptotic marker. Although the participation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service