Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MAK219

Sigma-Aldrich

Tyrosine Colorimetric Assay Kit

Sufficient for 100 Colorimetric tests

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12161503
NACRES:
NA.84

detection method

colorimetric

relevant disease(s)

neurological disorders

storage temp.

−20°C

General description

Tyrosine (Tyr) is an aromatic amino acid that is a precursor of several important substrates such as catecholamine, thyroid hormone, and melanin pigment. A deficiency in enzymes such as homogentisic acid oxidase or phenylalanine 4-hydroxylase that degrade tyrosine result in rare, hereditary metabolic disorders.

Suitability

Suitable for the detection of tyrosine in a variety of biological samples.

Principle

The Tyrosine Colorimetric Assay Kit provides a simple assay for measuring tyrosine in biological samples (ranging from 5-75 nmole/well). Tyrosine is determined by measuring a colorimetric product with absorbance at 492 nm (A492) proportional to the amount of tyrosine present.

Kit Components Only

Product No.
Description

  • Tyr Assay Buffer

  • Tyr Enzyme Mix

  • Tyr Standard

Pictograms

CorrosionHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li-Mao et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 84, 1923-1929 (2016-11-17)
To explore the new mechanism of neuroprtection of monosialotetrahexosylganglioside and providing reliable theoretical foundation and experimental evidence for the emergency treatment and rehabilitation of cerebral ischemia/reperfusion injury. A rat model of cerebral ischemia/reperfusion injury was constructed and intervened with monosialotetrahexosylganglioside(5mg/kg)
Delong Jiao et al.
Bioactive materials, 6(7), 2011-2028 (2021-01-12)
Developmental engineering strategy needs the biomimetic composites that can integrate the progenitor cells, biomaterial matrices and bioactive signals to mimic the natural bone healing process for faster healing and reconstruction of segmental bone defects. We prepared the gelatin-reduced graphene oxide
Li Mao et al.
Oncotarget, 8(59), 99681-99692 (2017-12-17)
MicroRNAs (miRNAs, miR) play a fundamental role in cerebral ischemia/reperfusion (I/R) injury. However, the role of miRNAs in toxic aldehyde and tyrosine accumulation is not fully elucidated. We constructed a cerebral I/R rat model and found that overexpression of miR-193

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service