Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE28-9042-50

CY®5,5 Amidite 100mg

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106000
NACRES:
NA.32

form

solid

manufacturer/tradename

Cytiva

solubility

soluble (Soluble in cold water and hot water)

General description

Produces exceptional sequencing results from difficult templates such as those with high GC content and secondary structures.

TempliPhi™ Sequence Resolver Kit resolves most of the sequencing stops or gaps that routinely elude other commonly used finishing methods. The use of this kit eliminates the need to use dGTP chemistry, which can affect the accuracy of a sequence due to the appearance of sequence compression. In addition, it eliminates the need for additives in the sequencing reaction.

Amersham™ CyDye™ fluors are versatile fluorophores designed for a broad range of applications. The use of multicolor CyDye fluors enables multiplexing without crosstalk. In addition, the CyDye flours readily dissolve in aqueous buffers, which eliminates the need of organic solvents. The high purity and thorough QC testing of Amersham CyDye fluors generate high levels of chromophore and reactive dye content. Narrow excitation and emission bands result in discrete signals from each fluor, which together with minimal cross-talk contributes to high accuracy.

Features and Benefits

  • Wide selection of CyDye fluors for protein labeling for use in ELISA & immunoprecipitation.
  • Labeling strategy based on protein characteristics.
  • Medium to high-throughput fluorescence imaging.
  • Ready-to-use labeled antibodies enable multiplexed assays.
  • Streptavidin-conjugated CyDye fluors for use in Flow-cytometry and In situ hybridization.
  • For incorporation of Cy5.5 fluorescent dyes at any position in an oligonucleotide during synthesis.
  • DNA synthesis reagent.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Other Notes

Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, away from incompatible materials. Keep container tightly closed and sealed until ready for use

Legal Information

Cy is a registered trademark of Cytiva

Pictograms

Exclamation mark

Signal Word

Danger

Storage Class Code

13 - Non Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service