Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

CWA-1164

Sigma-Aldrich

Anti-Human CD79a (HM57) ColorWheel® Dye-Ready mAb

greener alternative

Synonym(s):

B-cell antigen receptor complex-associated protein alpha chain;Ig-alpha;MB-1 membrane glycoprotein;Membrane-bound immunoglobulin-associated protein;Surface IgM-associated protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

HM57, monoclonal

product line

ColorWheel®

form

lyophilized

mol wt

calculated mol wt 25.04 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

chicken, rabbit, nonhuman primates, horse, rat, mouse, guinea pig

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

technique(s)

flow cytometry: suitable

isotype

IgG1

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

compatibility

for use with ColorWheel® Dyes (Required, (Sold Separately))

greener alternative category

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CD79A(973)

General description

ColorWheel® technology is a novel and proprietary method of creating your own antibody and dye combinations for use in flow cytometry. By incubating any ColorWheel® antibody with any ColorWheel® dye, researchers can quickly and simply produce primary conjugated antibodies for use in single color or multicolor flow cytometry analysis. Each ColorWheel® antibody and dye is lyophilized for long-term storage, allowing for simplicity and flexibility without compromise. ColorWheel® technology requires both ColorWheel® antibody and ColorWheel® dye for Flow Cytometry application.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This product is Preservative-free, lyophilized product for enhanced stability and allow for ambient shipping and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.

Specificity

Clone HM57 is a mouse monoclonal antibody that detects CD79a. It targets an epitope within the C-terminal, cytoplasmic domain.

Immunogen

A linear peptide corresponding to 15 amino acids from the C-terminal, cytoplasmic domain of human CD79a.

Application

Quality Control Testing

Evaluated by Flow Cytometry in Human peripheral blood mononuclear cells (PBMC), gated on lymphocytes.

Flow Cytometry Analysis: One million human peripheral blood mononuclear cells (PBMC) lymphocytes were stained with APC conjugated anti-human CD19, fixed, permeabilized and subsequently stained with 5 µL of a 1:1 mixture of Cat. No. CWA-1164, Anti-Human CD79a (HM57) ColorWheel® Dye-Ready mAb and Cat. No. CWDS-PE ColorWheel® Antibody-Ready Phycoerythrin (PE) Dye or an equivalent amount of Cat. No. CWA-1013 Mouse IgG1 isotype control and Cat. No. CWDS-PE ColorWheel® Antibody-Ready Phycoerythrin (PE) Dye. CD79a expression was observed in CD19 positive population.

Tested Applications

Flow Cytometry Analysis: One million human peripheral blood mononuclear cells (PBMC) lymphocytes were stained with PE conjugated anti-human CD19, fixed, permeabilized and subsequently stained with 5 µL of a 1:1 mixture of Cat. No. CWA-1164, Anti-Human CD79a (HM57) ColorWheel® Dye-Ready mAb and Cat. No. CWDS-APC ColorWheel® Antibody-Ready Allophycocyanin (APC) Dye or an equivalent amount of Cat. No. CWA-1013 Mouse IgG1 isotype control and Cat. No. CWDS-APC ColorWheel® Antibody-Ready Allophycocyanin (APC) Dye. CD79a expression was observed in CD19 positive population.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Compatibility

Requires ColorWheel® Dye (Sold Separately)

Target description

B-cell antigen receptor complex-associated protein alpha chain (UniProt: P11912; also known as Ig-alpha, MB-1 membrane glycoprotein, Membrane-bound immunoglobulin-associated protein, Surface IgM-associated protein, CD79a) is encoded by the CD79A (also known as IGA, MB1) gene (Gene ID: 973) in human. CD79a is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-32), which is subsequently cleaved off to produce the mature form that contains an extracellular domain (aa 33-143), a transmembrane domain (aa 144-165), and a cytoplasmic domain (aa 166-226). CD79a is required in cooperation with CD79b for initiation of the signal transduction cascade activated by binding of antigen to the B-cell antigen receptor complex (BCR) that leads to internalization of the complex, trafficking to late endosomes, and antigen presentation. It is also shown to be essential for BCR surface expression and for efficient differentiation of pro- and pre-B-cells. CD79a undergoes phosphorylation on tyrosine, serine, and threonine residues upon B-cell activation. Phosphorylation of tyrosine residues by Src-family kinases is an early and essential feature of the BCR signaling cascade. The phosphorylated tyrosines serve as docking sites for SH2-domain containing kinases, leading to their activation that in turn leads to phosphorylation of downstream targets. Phosphorylation of serine and threonine residues may prevent subsequent tyrosine phosphorylation. CD79a contains an ITAM domain that is localized in amino acids 107-205. Mutations in CD79A gene is known to cause autosomal recessive agammaglobulinemia 3, a primary immunodeficiency characterized by low or absent serum antibodies and low or absent circulating B-cells due to an early block of B-cell development. (Ref.: Rosenlow, J., et al. (2014). PLoS One. 9(4); e96199; Muller, R., et al. (2000). Proc. Natl. Acad. Sci. USA. 97(15); 8451-8454; Minegishi, Y., et al. (1999). J. Clin. Invest. 104(8); 1115-1121).

Physical form

Lyophilized from PBS containing D-Mannitol and Sucrose, normal appearance is a dried pellet. Reconstituted antibody solution is stable and functional as assessed by functional testing. Contains no biocide or preservatives, such as azide.

Reconstitution

1.0 mg/mL after reconstitution at 25 µL in PBS. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C. Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial. Reconstitute each vial by adding 25 µL PBS. Protect from light.

Legal Information

ColorWheel is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service