QC1275
PCBs in Transformer Oil
certified reference material
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
certified reference material
CofA
current certificate can be downloaded
packaging
ampule of 2 mL
manufacturer/tradename
RTC QCO-072
concentration
100% in mineral oil
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
environmental
format
matrix material
storage temp.
2-30°C
General description
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Application
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Other Notes
Values of analytes vary lot to lot. This product will contain one of the Aroclors from the Analyte table below.
Analyte
Description
Aroclor-1016 (PCB-1016) 10-50 mg/kg
Aroclor-1221 (PCB-1221) 12-50 mg/kg
Aroclor-1232 (PCB-1232) 12-50 mg/kg
Aroclor-1242 (PCB-1242) 10-50 mg/kg
Aroclor-1248 (PCB-1248) 12-50 mg/kg
Aroclor-1254 (PCB-1254) 10-50 mg/kg
Aroclor-1260 (PCB-1260) 10-50 mg/kg
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Asp. Tox. 1 - STOT RE 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service