Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

77518

Supelco

Nomilin

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H34O9
CAS Number:
Molecular Weight:
514.56
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤5.0% water

application(s)

food and beverages

format

neat

SMILES string

CC(=O)OC1CC(=O)OC(C)(C)[C@@H]2CC(=O)[C@]3(C)[C@H](CC[C@@]4(C)[C@@H](OC(=O)[C@H]5O[C@@]345)c6ccoc6)[C@@]12C

InChI

1S/C28H34O9/c1-14(29)34-19-12-20(31)36-24(2,3)17-11-18(30)27(6)16(26(17,19)5)7-9-25(4)21(15-8-10-33-13-15)35-23(32)22-28(25,27)37-22/h8,10,13,16-17,19,21-22H,7,9,11-12H2,1-6H3/t16-,17+,19?,21+,22-,25+,26-,27+,28-/m1/s1

InChI key

KPDOJFFZKAUIOE-BIYZDVJRSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinhee Kim et al.
European journal of pharmacology, 811, 93-100 (2017-05-30)
Nomilin is a bitter compound present in citrus and has been demonstrated as useful for various disease preventions through anti-proliferative, anti-inflammatory, and pro-apoptotic activities. Although in vitro disease models have shown that certain limonoids in the p38 mitogen-activated protein kinase
Seong Yeong Kim et al.
Preventive nutrition and food science, 18(3), 196-202 (2014-01-29)
Citron seed extracts (CSEs) were made using distilled water (CSEW), ethanol (CSEE), and n-hexane (CSEH), to measure the total polyphenol contents, DPPH and ABTS radical scavenging activities, and anti-complementary activity. The total polyphenol content was observed the highest in CSEE
Smita Raithore et al.
Sensors (Basel, Switzerland), 15(12), 30062-30075 (2015-12-04)
In an earlier study, an electronic tongue system (e-tongue) has been used to differentiate between orange juice made from healthy fruit and from fruit affected by the citrus greening or Huanglongbing (HLB) disease. This study investigated the reaction of an
Fuhua Yan et al.
PloS one, 13(7), e0200320-e0200320 (2018-07-10)
Carotenoids in citrus fruits have health benefits and make the fruits visually attractive. Red-fleshed 'Chuhong' ('CH') and pale green-fleshed 'Feicui' ('FC') pummelo (Citrus maxima (Burm) Merr.) fruits are interesting materials for studying the mechanisms of carotenoid accumulation. In this study
Xiaoyong Zheng et al.
Molecules (Basel, Switzerland), 24(7) (2019-04-13)
Gomisin D, a lignan compound isolated from Fructus Schisandra, is a potential antidiabetic and anti-Alzheimer's agent. Recently, gomisin D was used as a quality marker of some traditional Chinese medicine (TCM) formulas. In this study, a rapid ultra-performance liquid chromatography/tandem

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service