Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

36904

Supelco

PCB No 153 solution

10 μg/mL in isooctane, analytical standard

Synonym(s):

2,2′,4,4′,5,5′-Hexachlorobiphenyl solution, 2,2′,4,4′,5,5′-PCB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H4Cl6
CAS Number:
Molecular Weight:
360.88
Beilstein:
1990497
Ballschmiter Number:
153
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

10 μg/mL in isooctane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

SMILES string

Clc1cc(Cl)c(cc1Cl)-c2cc(Cl)c(Cl)cc2Cl

InChI

1S/C12H4Cl6/c13-7-3-11(17)9(15)1-5(7)6-2-10(16)12(18)4-8(6)14/h1-4H

InChI key

MVWHGTYKUMDIHL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cristina L Quinn et al.
Environment international, 49, 83-91 (2012-09-18)
Concentrations of persistent organic pollutants (POPs) in Inuit populations have been observed to decrease over the last decade. The main objective of this study was to develop a methodology to quantify the potential influence of intergenerational dietary transitions on human
Xue Shi et al.
Journal of proteome research, 11(7), 3805-3815 (2012-06-13)
Polychlorinated biphenyls (PCBs) are persistent organic pollutants and have been associated with abnormal liver enzymes and suspected nonalcoholic fatty liver disease (NAFLD), obesity, and the metabolic syndrome in epidemiological studies. In epidemiological surveys of human PCB exposure, PCB 153 has
Lei Chen et al.
Environmental science and pollution research international, 20(3), 1842-1848 (2012-09-06)
The ubiquitous dissolved organic matter (DOM) has an important influence on transformation of organic contaminants through the production of reactive substances, such as (•)OH, (1)O(2), and (3)DOM*. The photolysis of a higher chlorinated polychlorinated biphenyl (PCB) congener (2,2',4,4',5,5'-hexachlorobiphenyl, PCB 153)
Helen Engelstad Kvalem et al.
Environment international, 50, 15-21 (2012-10-04)
Dioxins and PCBs accumulate in the food chain and might exert toxic effects in animals and humans. In large epidemiologic studies, exposure estimates of these compounds based on analyses of biological material might not be available or affordable. To develop
Margherita Calò et al.
Environmental toxicology and pharmacology, 34(3), 735-742 (2012-11-06)
The effects of polychlorinated biphenyl congeners 126 and 153 (PCB-126 and PCB-153) on vitellogenin (Vtg) and cytochrome (CYP1A1) expression were evaluated in 60 juvenile Sparus aurata. Fish were divided into four groups and the control group (Group A) was compared

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service