Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

13-1885

Sigma-Aldrich

Hydrofluoric acid

SAJ first grade

Synonym(s):

HF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HF
CAS Number:
Molecular Weight:
20.01
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

SAJ first grade

vapor density

1.27 (vs air)

vapor pressure

25 mmHg ( 20 °C)

form

liquid

availability

available only in Japan

density

1.15 g/mL at 25 °C (lit.)

SMILES string

[H+].[F-]

InChI

1S/FH/h1H

InChI key

KRHYYFGTRYWZRS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ian Ford et al.
International journal of cardiology, 184, 163-169 (2015-02-24)
We identified easily obtained baseline characteristics associated with outcomes in patients with chronic heart failure (HF) and elevated heart rate (HR) receiving contemporary guideline-recommended therapy in the SHIFT trial, and used them to develop a prognostic model. We selected the
Wenlin Yvonne Lin et al.
Waste management (New York, N.Y.), 41, 75-84 (2015-04-22)
Accelerated carbonation has been studied as a treatment method for MSW IBA, and the main advantage is that it can shorten the treatment duration from months to days, compared to natural weathering. This study investigated the effect of accelerated carbonation
Yvonne C J Wientjes et al.
Genetics, selection, evolution : GSE, 47, 42-42 (2015-05-09)
Although simulation studies show that combining multiple breeds in one reference population increases accuracy of genomic prediction, this is not always confirmed in empirical studies. This discrepancy might be due to the assumptions on quantitative trait loci (QTL) properties applied
Faiez Zannad et al.
European journal of heart failure, 17(7), 735-742 (2015-04-29)
Thrombin is a critical element of crosstalk between pathways contributing to worsening of established heart failure (HF). The aim of this study is to explore the efficacy and safety of rivaroxaban 2.5 mg bid compared with placebo (with standard care) after
Barry Greenberg et al.
JACC. Heart failure, 2(1), 84-92 (2014-03-14)
Impaired cardiac isoform of sarco(endo)plasmic reticulum Ca(2+) ATPase (SERCA2a) activity is a key abnormality in heart failure patients with reduced ejection fraction. The CUPID 2 (Calcium Up-Regulation by Percutaneous Administration of Gene Therapy in Cardiac Disease Phase 2b) trial is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service