Key Documents
PA12/111
Millipore
NovAseptic® Valve, Pneumatic Plastic Actuator, Normally Closed, NC
About This Item
UNSPSC Code:
40141600
eCl@ss:
37051001
material
POM-C indicator
cover (Stainless Steel EN 1.4305/ASTM 303 and Plastic POM C)
Quality Level
product line
NovAseptic®
parameter
pressure range at -1--6 bar
temp. range at 0-70 °C
weight
0.3 kg (0.66 lb)
Application
The applied valve body and diaphragm may have different design temperature and/or pressure limits. The weakest component in the assembled product determines the maximum design temperature and pressure limits.
Other Notes
Actuator, Pneumatic
Legal Information
NOVASEPTIC is a registered trademark of Merck KGaA, Darmstadt, Germany