Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABD105

Sigma-Aldrich

Anti-Wnt-11 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

WNT11

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

bovine (based on 100% sequence homology), mouse (based on 100% sequence homology), rat (based on 100% sequence homology), rabbit (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... WNT11(7481)

General description

Wnt genes belong to a family of protooncogenes with at least 13 known members that are expressed in species ranging from Drosophila to man. Transcription of Wnt family genes appears to be developmentally regulated in a precise temporal and spatial manner. Wnt-11 activity is required for cells to undergo correct convergent extension movements during gastrulation. Wnt-11 is also expressed in the lung mesenchyme, the urorectal septum, the urogenital folds, the labioscrotal swellings, and the cortex of the adrenal gland. Unlike other Wnt family members, Wnt-11 is not expressed in the neuroepithelium of the central nervous system. Wnt-11, along with Wnt-8c, is expressed in the posterior region of the chick embryo in the caudal paraxial mesoderm that underlies the prospective caudal neural plate.

Immunogen

KLH-conjugated linear peptide corresponding to the C-terminus of Human Wnt-11.

Application

This Anti-Wnt-11 Antibody is validated for use in Western Blotting for the detection of Wnt-11.

Quality

Evaluated by Western Blot in human osteosarcoma U2OS cell lysate.
Western Blot Analysis: 0.5 µg/mL of this antibody detected WNT-11 in 10 µg of human osteosarcoma U2OS cell lysate.

Target description

~39 kDa observed

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service