Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

6.18602

Supelco

Cellulose microcrystal

powder, for TLC

Synonym(s):

Microcrystalline Cellulose

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352201

Quality Level

form

powder

Application


  • Evaluating the effects of water matrix constituents on micropollutant removal by activated carbon and β-cyclodextrin polymer adsorbents.: Research focusing on water treatment, where cellulose microcrystals could play a role in enhancing the efficacy of micropollutant adsorption from various water matrices (Ling et al., 2020).

  • Fabrication of Cellulose Nanocrystal/Silver/Alginate Bionanocomposite Films with Enhanced Mechanical and Barrier Properties for Food Packaging Application.: This paper discusses the use of cellulose microcrystals in creating bionanocomposite films for food packaging, improving mechanical strength and barrier properties (Yadav et al., 2019).

  • Dissolution of cellulose in ionic liquid and water mixtures as revealed by molecular dynamics simulations.: Offers insights into the solvation processes of cellulose in ionic liquids, potentially enhancing its applications in biomaterials and nanocomposites (Manna & Ghosh, 2019).

  • Interfacial Engineering of Silicon Carbide Nanowire/Cellulose Microcrystal Paper toward High Thermal Conductivity.: Investigates the interfacial properties of silicon carbide nanowire/cellulose microcrystal composites, aiming to improve their thermal conductivity for advanced materials applications (Yao et al., 2016).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service