Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

572632-M

Millipore

SU5614

A potent, cell-permeable, reversible, ATP-competitive, and selective inhibitor of VEGF (Flk-1; IC₅₀ = 1.2 µM) and PDGF (IC₅₀ = 2.9 µM) receptor tyrosine kinases.

Synonym(s):

SU5614, 5-Chloro-3-[(3,5-dimethylpyrrol-2-yl)methylene]-2-indolinone, VEGFR Tyrosine Kinase Inhibitor VII, VEGFR2 Kinase Inhibitor IX

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13ClN2O
CAS Number:
Molecular Weight:
272.73
UNSPSC Code:
12352200

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange-red

solubility

DMSO: 10 mg/mL

storage temp.

2-8°C

InChI

1S/C15H13ClN2O/c1-8-5-9(2)17-14(8)7-12-11-6-10(16)3-4-13(11)18-15(12)19/h3-7,17H,1-2H3,(H,18,19)

InChI key

XLBQNZICMYZIQT-UHFFFAOYSA-N

General description

A potent, cell-permeable, reversible, ATP-competitive, and selective inhibitor of VEGF (Flk-1; IC50 = 1.2 µM) and PDGF (IC50 = 2.9 µM) receptor tyrosine kinases. Does not have any effect on the EGF and IGF receptor tyrosine kinases. Also inhibits the VEGF-driven mitogenesis of human umbilical vein endothelial cells (HUVECs; IC50 ≤680 nM). Inhibits Flt3 phosphorylation in Flt3/Itd-BaF3 cells.
A potent, cell-permeable, reversible, ATP-competitive, and selective inhibitor of VEGF (Flk-1; IC50 = 1.2 µM) and PDGF (IC50 = 2.9 µM) receptor tyrosine kinases. Does not have any effect on the EGF and IGF receptor tyrosine kinases. Also inhibits the VEGF-driven mitogenesis of human umbilical vein endothelial cells (HUVECs; IC50 ≤ 680 nM). Inhibits Flt3 phosphorylation in Flt3/Itd-BaF3 cells.

Biochem/physiol Actions

Primary Target
VEGF Flk-1
Target IC50: 1.2 µM, 2.9 µM against VEGF (Flk-1) and PDGF receptor tyrosine kinases, respectively; ≤680 nM against the VEGF-driven mitogenesis of human umbilical vein endothelial cells (HUVECs)

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 2 months at -20°C.

Other Notes

Cools, J., et al. 2004. Cancer Res.64, 6385.
Sun, L., et al. 1998. J. Med. Chem. 41, 2588.

Legal Information

Not available for sale in the United States.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service