Recommended Products
technique(s)
photometry: suitable
Quality Level
compatibility
for use with Spectroquant®
storage temp.
2-30°C
General description
This absorption tube is a required accessory when performing the Spectroquant® COD cell tests for seawater or other samples with high chloride content (117058 and 117059). The chloride content of the water sample is depleted by means of sulfuric acid and an HCl absorber. The chloride depletion method corresponds to DIN 38409-41-2.
Legal Information
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service