50690
Glyoxal bis(sodium hydrogen sulfite) adduct hydrate
technical, ~90% (T)
Synonym(s):
1,2-Dihydroxy-1,2-ethanedisulfonic acid disodium salt
Sign Into View Organizational & Contract Pricing
Select a Size
All Photos(1)
Select a Size
Change View
About This Item
Linear Formula:
OHCCHO · 2NaHSO3
CAS Number:
Molecular Weight:
266.16 (anhydrous basis)
Beilstein:
3898061
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
grade
technical
Quality Level
Assay
~90% (T)
form
solid
impurities
~10% water
functional group
sulfonic acid
SMILES string
O.[Na+].[Na+].OC(C(O)S([O-])(=O)=O)S([O-])(=O)=O
InChI
1S/C2H6O8S2.2Na.H2O/c3-1(11(5,6)7)2(4)12(8,9)10;;;/h1-4H,(H,5,6,7)(H,8,9,10);;;1H2/q;2*+1;/p-2
InChI key
JQYGKJGYGOFFIK-UHFFFAOYSA-L
General description
Glyoxal bis(sodium hydrogen sulfite) adduct hydrate is a gyloxal derivative.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service