Key Documents
Z516813
Sigma-Aldrich
Aldrich® three-neck round-bottom flask
vertical, capacity 1,000 mL, joint: ST/NS 29/32
Sinônimo(s):
flask round bottom, round bottom flasks, round bottomed flask
About This Item
Código UNSPSC:
41121804
NACRES:
NB.43
Materiais
borosilicate glass
round bottom flask
three-neck (Vertical)
three-neck
Junta
joint: ST/NS29/32
esterilidade
non-sterile
Características
closure type Standard Taper Joint
embalagem
pack of 1
fabricante/nome comercial
Sigma-Aldrich
capacidade
1,000 mL
Informações legais
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC