Key Documents
CLS362220
Pyrex® Barrett distilling apparatus, ST joints
capacity 20 mL
Sinônimo(s):
Corning® 3622
About This Item
Código UNSPSC:
41121800
NACRES:
NB.43
fabricante/nome comercial
Corning 3622-20
disponibilidade
not available in EU
capacidade
20 mL
Descrição geral
20 mL, with 24/40 joints and ST No.1 stopcock plug.
Informações legais
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.