Merck
Possible involvement of chemokine C-C receptor 7
Journal of gastroenterology and hepatology (2017-06-08)
Naruhiro Kimura, Satoshi Yamagiwa, Tomoyuki Sugano, Toru Setsu, Kentaro Tominaga, Hiroteru Kamimura, Masaaki Takamura, Shuji Terai
ABSTRACT
Recent studies have demonstrated that B cells and follicular helper T (Tfh) cells, which are central regulators of humoral immune response, contribute to the development and progression of autoimmune diseases. Because Tfh cells can be divided into several subsets with distinct functional properties, this study aimed to examine the roles of different subsets of circulating Tfh cells in the immune pathogenesis of autoimmune hepatitis (AIH). Thirty-five patients with AIH, 28 patients with primary biliary cholangitis, 22 patients with chronic hepatitis B (CHB), and 44 health controls (HC) were enrolled. The frequencies of different Tfh subsets in the blood and liver were examined by flow cytometry and immunohistochemical staining. The function of circulating Tfh subsets was examined after in vitro stimulation. In newly diagnosed AIH patients, the frequency of circulating chemokine C-C receptor 7 These results significantly extend our understanding of Tfh subsets in AIH and suggest a potential role of dysregulated chemokine C-C receptor 7