- Obesity-promoting and anti-thermogenic effects of neutrophil gelatinase-associated lipocalin in mice.
Obesity-promoting and anti-thermogenic effects of neutrophil gelatinase-associated lipocalin in mice.
Scientific reports (2017-11-16)
Akira Ishii, Goro Katsuura, Hirotaka Imamaki, Hiroyuki Kimura, Keita P Mori, Takashige Kuwabara, Masato Kasahara, Hideki Yokoi, Kousaku Ohinata, Tomoko Kawanishi, Junichi Tsuchida, Yuji Nakamoto, Kazuwa Nakao, Motoko Yanagita, Masashi Mukoyama, Kiyoshi Mori
PMID29138470
ABSTRACT
Neutrophil gelatinase-associated lipocalin (NGAL, lipocalin 2 or LCN2) is an iron carrier protein whose circulating level is increased by kidney injury, bacterial infection and obesity, but its metabolic consequence remains elusive. To study physiological role of LCN2 in energy homeostasis, we challenged female Lcn2 knockout (KO) and wild-type (WT) mice with high fat diet (HFD) or cold exposure. Under normal diet, physical constitutions of Lcn2 KO and WT mice were indistinguishable. During HFD treatment, Lcn2 KO mice exhibited larger brown adipose tissues (BAT), consumed more oxygen, ate more food and gained less body weights as compared to WT mice. When exposed to 4 °C, KO mice showed higher body temperature and more intense