- Impact of Aerosol Dust on xMAP Multiplex Detection of Different Class Pathogens.
Impact of Aerosol Dust on xMAP Multiplex Detection of Different Class Pathogens.
Frontiers in microbiology (2017-12-15)
Denis A Kleymenov, Vladimir A Gushchin, Alexander L Gintsburg, Artem P Tkachuk
PMID29238328
ABSTRACT
Environmental or city-scale bioaerosol surveillance can provide additional value for biodefense and public health. Efficient bioaerosol monitoring should rely on multiplex systems capable of detecting a wide range of biologically hazardous components potentially present in air (bacteria, viruses, toxins and allergens). xMAP technology from Luminex