Merck
  • miR-34a Targets HDAC1-Regulated H3K9 Acetylation on Lipid Accumulation Induced by Homocysteine in Foam Cells.
miR-34a Targets HDAC1-Regulated H3K9 Acetylation on Lipid Accumulation Induced by Homocysteine in Foam Cells.
Journal of cellular biochemistry (2017-05-10)
Qingguo Zhao, Shuqiang Li, Nan Li, Xiaoling Yang, Shengchao Ma, Anning Yang, Hui Zhang, Songhao Yang, Caiyan Mao, Lingbo Xu, Tingting Gao, Xiaoming Yang, Huiping Zhang, Yideng Jiang
ABSTRACT
Hyperhomocysteinemia (HHcy) promotes atherogenesis by modification of histone acetylation patterns and regulation of miRNA expression while the underlying molecular mechanisms are not well known. In this study, we investigated the effects of homocysteine (Hcy) on the expression of histone deacetylase 1 (HDAC1) and tested our hypothesis that Hcy-induced atherosclerosis is mediated by increased HDAC1 expression, which is regulated by miR-34a. The expression of HDAC1 increased and acetylation of histone H3 at lysine 9 (H3K9ac) decreased in the aorta of ApoE