Merck
  • B cell antigen receptor-induced activation of an IRAK4-dependent signaling pathway revealed by a MALT1-IRAK4 double knockout mouse model.
B cell antigen receptor-induced activation of an IRAK4-dependent signaling pathway revealed by a MALT1-IRAK4 double knockout mouse model.
Cell communication and signaling : CCS (2011-03-15)
Almut Dufner, Wolfgang W Schamel
ABSTRACT
The B cell antigen receptor (BCR) and pathogen recognition receptors, such as Toll-like receptor 4 (TLR4), act in concert to control adaptive B cell responses. However, little is known about the signaling pathways that integrate BCR activation with intrinsic TLR4 stimulation. Antigen receptors initialize activation of the inducible transcription factor nuclear factor-κB (NF-κB) via recruitment of the membrane-associated guanylate kinase caspase recruitment domain protein 11 (CARD11), the adapter molecule B cell CLL/lymphoma 10 (BCL10), and the "paracaspase" mucosa-associated lymphoid tissue lymphoma translocation gene 1 (MALT1) into lipid rafts. Upon BCR triggering, this activation strictly depends on BCL10, but not on MALT1, leading to the hypothesis that a MALT1-independent NF-κB activation pathway contributes to BCR-induced NF-κB activation downstream of BCL10. The identity of this pathway has remained elusive. Using genetic and biochemical approaches, we demonstrate that the IRAK4- and IRAK1-dependent TLR signaling branch is activated upon BCR triggering to induce partial NF-κB activation. BCR-induced MALT1-independent IκB degradation and B cell proliferation were inhibited in MALT1/IRAK4 double knockout B cells. Moreover, IRAK1 was recruited into lipid rafts upon BCR stimulation and activated following transient recruitment of IRAK4. We propose that the observed crosstalk between BCR and TLR signaling components may contribute to the discrimination of signals that emanate from single and dual receptor engagement to control adaptive B cell responses.