Merck
  • Copeptin levels associate with cardiovascular events in patients with ESRD and type 2 diabetes mellitus.
Copeptin levels associate with cardiovascular events in patients with ESRD and type 2 diabetes mellitus.
Journal of the American Society of Nephrology : JASN (2011-03-19)
Wiebke Fenske, Christoph Wanner, Bruno Allolio, Christiane Drechsler, Katja Blouin, Jürgen Lilienthal, Vera Krane
ABSTRACT
In ESRD, the neurohormone arginine vasopressin (AVP) may act primarily through V1a and V1b receptors, which promote vasoconstriction, myocardial hypertrophy, and release of adrenocorticotropic hormone. The preanalytical instability of AVP limits the investigation of whether this hormone associates with cardiovascular events, but the stable glycopeptide copeptin may serve as a surrogate because it is co-secreted with AVP from the posterior pituitary. Here, we studied whether copeptin predicts cardiovascular risk and mortality in ESRD. We measured copeptin at baseline in 1241 hemodialysis patients with type 2 diabetes participating in the German Diabetes and Dialysis Study. The median copeptin level was 81 pmol/L (interquartile range, 81 to 122 pmol/L). In Cox regression analyses, compared with patients with copeptin levels in the lowest quartile (≤51 pmol/L), patients with copeptin levels in the highest quartile (>122 pmol/L) had a 3.5-fold increased risk for stroke (HR, 3.48; 95% CI: 1.71 to 7.09), a 73% higher risk for sudden death (HR, 1.73; 95% CI: 1.01 to 2.95), a 42% higher risk for combined cardiovascular events (HR, 1.42; 95% CI: 1.06 to 1.90), and a 48% higher risk for all-cause mortality (HR, 1.48; 95% CI: 1.15 to 1.90). In contrast, we did not detect significant associations between copeptin levels and risks for myocardial infarction or death caused by congestive heart failure. In conclusion, copeptin levels strongly associate with stroke, sudden death, combined cardiovascular events, and mortality in hemodialysis patients with type 2 diabetes. Whether vasopressin receptor antagonists will improve these outcomes requires further studies.