Merck
  • Hepatitis B virus X stimulates redox signaling through activation of ataxia telangiectasia mutated kinase.
Hepatitis B virus X stimulates redox signaling through activation of ataxia telangiectasia mutated kinase.
International journal of clinical and experimental pathology (2014-06-27)
Yasunobu Matsuda, Ayumi Sanpei, Toshifumi Wakai, Masayuki Kubota, Mami Osawa, Yuki Hirose, Jun Sakata, Takashi Kobayashi, Shun Fujimaki, Masaaki Takamura, Satoshi Yamagiwa, Masahiko Yano, Shogo Ohkoshi, Yutaka Aoyagi
ABSTRACT
Hepatitis B virus X (HBX) protein plays a crucial role in carcinogenesis, but its mechanism is unclear. The involvement of ataxia telangiectasia mutated (ATM) kinase in the enhanced redox system was investigated by examining the phosphorylation level of ATM in HBX gene-transfected cells and in transgenic mice following redox system manipulation by treatment with hydrogen peroxide (H2O2) or antioxidant. Western blotting and immunostaining showed that phospho-ATM was significantly increased by HBX both in vitro (3.2-fold; p<0.05) and in vivo (4-fold; p<0.05), and this effect was abrogated by antioxidant treatment. The level of PKC-δ in HBX-expressing cells was increased 3.5-fold compared to controls. Nuclear localized NF-E2-related factor 2 (Nrf2) was increased in HBX-expressing cells exposed to H2O2, but remained at lower levels after the treatment with rottlerin, KU55933, or caffeine. The levels of anti-oxidant molecules were increased in HBX expressing cells and in transgenic mice, indicating that HBX stimulates the Nrf2-mediated redox system. The levels of intracellular reactive oxygen species (ROS) were significantly increased in HBX-expressing cells treated with hydrogen peroxide in the presence of ATM inhibitor KU55933 or caffeine. Treatment of HBX-expressing cells with KU55933 or caffeine before the exposure to H2O2 increased the ratio of cell apoptosis to 33±4% (p<0.05) and 22±4% (p<0.05), respectively. Collectively, HBX stimulates the ATM-mediated PKC-δ/Nrf2 pathway, and maintains the enhanced activity of the redox system. Therefore, manipulating ATM kinase activity might be a useful strategy for treating HBX-induced carcinogenesis.
Product Description
Supelco
p-Benzoquinone, for spectrophotometric det. of amines, ≥99.5% (HPLC)
Supelco
1,4-Benzoquinone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Melting point standard 235-237°C, analytical standard
Sigma-Aldrich
p-Benzoquinone, reagent grade, ≥98%
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Caffeine, European Pharmacopoeia (EP) Reference Standard
Supelco
Caffeine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material