Merck
  • Entomotoxicity and biosafety assessment of PEGylated acephate nanoparticles: a biologically safe alternative to neurotoxic pesticides.
Entomotoxicity and biosafety assessment of PEGylated acephate nanoparticles: a biologically safe alternative to neurotoxic pesticides.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2013-04-16)
Saheli Pradhan, Indrani Roy, Gopal Lodh, Prasun Patra, Samrat Roy Choudhury, Arunava Samanta, Arunava Goswami
ABSTRACT
This is a report of an experimental study on a nanoencapsulation of the organophosphate acephate. Acephate was encapsulated in polyethylene glycol, using a simple, easy-to-replicate method that required no special equipment or conditions. The nanoencapsulation (nanoacephate) was characterized and its bioefficacy as compared to the regular commercial acephate was tested. The biosafety of the new compound was also tested on a murine model. Our new nanoencapsulation scored over the regular variety on all counts. It was found to successfully incorporate the active pesticidal component, acephate and this compound retained greater functional integrity over time as a nanoencapsulation. It was significantly more efficacious than the regular variety. It was biosafe when tested on murine model. We have reason to believe that this nanoencapsulation would allow the use of an organophosphate in a more targeted manner, thereby making it a cost-effective and eco-friendly alternative to the regular variety in use now.