Merck
  • The clozapine metabolite N-desmethylclozapine displays variable activity in diverse functional assays at human dopamine D₂ and serotonin 5-HT₁A receptors.
The clozapine metabolite N-desmethylclozapine displays variable activity in diverse functional assays at human dopamine D₂ and serotonin 5-HT₁A receptors.
European journal of pharmacology (2011-08-13)
Peter Heusler, Liesbeth Bruins Slot, Amélie Tourette, Stéphanie Tardif, Didier Cussac
ABSTRACT
N-desmethylclozapine (NDMC or norclozapine) is the major active metabolite of the antipsychotic clozapine in humans. The activity of NDMC differs from clozapine at a number of neurotransmitter receptors, probably influencing the pharmacological effects of clozapine treatment. Here, we tested the properties of NDMC in comparison with clozapine at recombinant human dopamine D(2) and serotonin 5-HT(1A) receptors, using a panel of functional assays implicating diverse signalling pathways. At dopamine D(2) receptors, NDMC as well as clozapine did not display agonist activity in measures of G protein activation by [(35)S]GTPγS binding and in the sensitive Extracellular Signal-Regulated Kinase 1/2 (ERK1/2) phosphorylation assay. In contrast, there were weak partial agonist actions of NDMC (but not of clozapine) for dopamine D(2)-dependent activation of Ca(2+) liberation via coexpressed chimeric Gα(q/o) proteins and for G protein-coupled inward rectifier potassium channel (GIRK) current induction in Xenopus oocytes. Intriguingly, GIRK currents induced by NDMC via dopamine D(2) receptors showed a rapid and transient time course, strikingly different from currents recorded with other receptor agonists. At serotonin 5-HT(1A) receptors, NDMC was a more efficacious partial agonist than clozapine for [(35)S]GTPγS binding, ERK1/2 phosphorylation and GIRK activation. Respective low and moderate partial agonist properties of NDMC at dopamine D(2) and serotonin 5-HT(1A) receptors thus differentiate the metabolite from its parent drug and may contribute to the overall effects of clozapine pharmacotherapy.
Product Description
Supelco
N-Desmethylclozapine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®