Merck
  • N-Methylimidazole-catalyzed synthesis of carbamates from hydroxamic acids via the Lossen rearrangement.
N-Methylimidazole-catalyzed synthesis of carbamates from hydroxamic acids via the Lossen rearrangement.
Organic letters (2013-01-19)
Sabesan Yoganathan, Scott J Miller
ABSTRACT
An efficient, one-pot, N-methylimidazole (NMI) accelerated synthesis of aromatic and aliphatic carbamates via the Lossen rearrangement is reported. NMI is a catalyst for the conversion of isocyanate intermediates to the carbamates. Moreover, the utility of arylsulfonyl chloride in combination with NMI minimizes the formation of often-observed hydroxamate-isocyanate dimers during the sequence. Under the present conditions, lowering of temperatures is also possible, enabling a mild protocol.
Sigma-Aldrich
1-Methylimidazole
Sigma-Aldrich
1-Methylimidazole, ReagentPlus®, 99%
Sigma-Aldrich
1-Methylimidazole, ≥99%, purified by redistillation