Merck
  • Dexamethasone Downregulates SLC7A5 Expression and Promotes Cell Cycle Arrest, Autophagy and Apoptosis in BeWo Cells.
Dexamethasone Downregulates SLC7A5 Expression and Promotes Cell Cycle Arrest, Autophagy and Apoptosis in BeWo Cells.
Journal of cellular physiology (2015-06-23)
Bin He, Nana Zhang, Ruqian Zhao
ABSTRACT
Synthetic glucocorticoids (GCs) such as dexamethasone (Dex) are widely given to pregnant women to induce maturation and improve viability of preterm infants. Despite the beneficial effects, synthetic GCs have adverse effects on placental growth and nutrient transport system. However, the molecular mechanisms involved in these events remain unknown. Here we use a human placental choriocarcinoma cell line (BeWo) as model to explore the pathway linking amino acids transport with cell viability under Dex challenge. BeWo cells treated with Dex (100 nM) for 24 h demonstrated G1/S cell cycle arrest together with enhanced autophagy and apoptosis. Concurrently, the amino acid carrier SLC7A5 was down-regulated in association with impaired cellular amino acids uptake and inhibition of mammalian target of rapamycin (mTOR) signaling. Similar cellular responses were observed in BeWo cells treated with BCH, a classical System L inhibitor which inactivates SLC7A5. The glucocorticoid receptor (GR) antagonist RU486 was able to diminish Dex-induced translocation of GR into nucleus and to abolish these effects. Furthermore, Dex treatment significantly promoted the binding of GR to the proximal promoter sequence of SLC7A5 gene. Taken together, our results show that Dex downregulates SLC7A5 expression via GR-mediated transrepression. The impaired amino acids uptake leads to inhibition of mTOR signaling which in turn causes inhibited proliferation and enhanced autophagy and apoptosis in BeWo cells. These findings indicate that SLC7A5 mediates the effect of Dex on cell viability, thus providing a novel molecular target for the prevention and treatment of Dex-induced cell cycle arrest and apoptosis in placental cells.
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Roche
cOmplete ULTRA Tablets, Mini, EDTA-free, EASYpack Protease Inhibitor Cocktail, Tablets supplied in foil blister packs.