Merck
Optimal enzymatic hydrolysis of urinary benzodiazepine conjugates.
Journal of analytical toxicology (1994-11-01)
R Meatherall
ABSTRACT
Conditions for the enzymatic hydrolysis of benzodiazepine conjugates were systematically examined. Optimal recovery of the free drugs occurs when 1 mL of urine buffered to pH 4.5 is incubated with 5000 U of Helix pomatia beta-glucuronidase at 56 degrees C for 2 h. Urine specimens containing conjugates of temazepam, oxazepam, lorazepam, alpha-hydroxyalprazolam, 2-hydroxyethylflurazepam, and N-desalkyl-3-hydroxyflurazepam as targeted benzodiazepines were used. The freed drugs were quantitated by gas chromatography-mass spectrometry.
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, Type IX-A, lyophilized powder, 1,000,000-5,000,000 units/g protein (30 min assay)