Merck
  • AKT1/PKBalpha is recruited to lipid rafts and activated downstream of PKC isotypes in CD3-induced T cell signaling.
AKT1/PKBalpha is recruited to lipid rafts and activated downstream of PKC isotypes in CD3-induced T cell signaling.
FEBS letters (2003-04-23)
Birgit Bauer, Marcel Jenny, Fiedrich Fresser, Florian Uberall, Gottfried Baier
ABSTRACT
Protein kinase (PK) Ctheta and Akt/PKBalpha cooperate in T cell receptor/CD28-induced T cell signaling. We here demonstrate the recruitment of endogenous Akt1 and PKCtheta to lipid rafts in CD3-stimulated T cells. Further we show that Myr-PKCtheta mediates translocation of endogenous Akt1 to the plasma membrane as well as to lipid rafts, most likely explained by the observed complex formation of both protein kinases. In addition, in peripheral mouse T cells, the PKC inhibitor Gö6850 could partially block Akt1 activation in CD3-induced signaling, placing PKC isotype(s) upstream of Akt1. However, T cells derived from PKCtheta knockout mice were not impaired in CD3- or phorbol ester-induced Akt1 activity. Taken together, the results of this study give new insights into the functional link of Akt1 and PKCtheta in T cell signaling, demonstrating the co-recruitment of the two kinases and showing a novel pathway leading to Akt1 transactivation where PKC isotype(s) are involved but PKCtheta is not essential.