- Corrigendum to "Inhibition of cAMP/PKA Pathway Protects Optic Nerve Head Astrocytes against Oxidative Stress by Akt/Bax Phosphorylation-Mediated Mfn1/2 Oligomerization".
Corrigendum to "Inhibition of cAMP/PKA Pathway Protects Optic Nerve Head Astrocytes against Oxidative Stress by Akt/Bax Phosphorylation-Mediated Mfn1/2 Oligomerization".
Oxidative medicine and cellular longevity (2020-10-22)
Won-Kyu Ju, Myoung Sup Shim, Keun-Young Kim, Tae Lim Park, Sangphil Ahn, Genea Edwards, Robert N Weinreb
PMID33082914
ABSTRACT
[This corrects the article DOI: 10.1155/2019/8060962.].