Merck
  • CHIP Is an Essential Determinant of Neuronal Mitochondrial Stress Signaling.
CHIP Is an Essential Determinant of Neuronal Mitochondrial Stress Signaling.
Antioxidants & redox signaling (2015-01-21)
Amy M Palubinsky, Jeannette N Stankowski, Alixandra C Kale, Simona G Codreanu, Robert J Singer, Daniel C Liebler, Gregg D Stanwood, BethAnn McLaughlin
ABSTRACT
Determine the mechanism by which C-terminus of HSC70-interacting protein (CHIP) induction alters neuronal survival under conditions of mitochondrial stress induced by oxygen glucose deprivation. We report that animals deficient in the E3 ubiquitin ligase, CHIP, have high baseline levels of central nervous system protein oxidation and lipid peroxidation, reduced antioxidant defenses, and decreased energetic status. Stress-associated molecules typically linked to Parkinson's disease such as the mitochondrial kinase, PTEN-inducible putative kinase 1 (PINK1), and another E3 ligase, Parkin, are upregulated in brains from CHIP knockout (KO) animals. Utilizing a novel biotin-avidin capture technique, we found that the oxidation status of Parkin and the mitochondrial fission protein, dynamin-related protein 1 (Drp1), are altered in a CHIP-dependent manner. We also found that following oxygen-glucose deprivation (OGD), the expression of CHIP, PINK1, and the autophagic marker, LC3, increase and there is activation of the redox-sensitive kinase p66(shc). Under conditions of OGD, CHIP relocalizes from the cytosol to mitochondria. Mitochondria from CHIP KO mice have profound impairments in stress response induced by calcium overload, resulting in accelerated permeability transition activity. While CHIP-deficient neurons are morphologically intact, they are more susceptible to OGD consistent with a previously unknown neuroprotective role for CHIP in maintaining mitochondrial homeostasis. CHIP relocalization to the mitochondria is essential for the regulation of mitochondrial integrity and neuronal survival following OGD. CHIP is an essential regulator of neuronal bioenergetics and redox tone. Altering the expression of this protein has profound effects on neuronal survival when cells are exposed to OGD.
Sigma-Aldrich
In Vitro Toxicology Assay Kit, Lactic Dehydrogenase based
Sigma-Aldrich
Anti-CHIP Rabbit pAb, liquid, Calbiochem®
Sigma-Aldrich
OxyBlot Protein Oxidation Detection Kit, The OxyBlot Protein Oxidation Detection Kit provides the reagents to perform the immunoblot detection of carbonyl groups introduced into proteins by oxidative reactions with ozone or oxides of nitrogen or by metal catalyzed oxidation.