Merck
  • The Arabidopsis SAFEGUARD1 suppresses singlet oxygen-induced stress responses by protecting grana margins.
The Arabidopsis SAFEGUARD1 suppresses singlet oxygen-induced stress responses by protecting grana margins.
Proceedings of the National Academy of Sciences of the United States of America (2020-03-13)
Liangsheng Wang, Dario Leister, Li Guan, Yi Zheng, Katja Schneider, Martin Lehmann, Klaus Apel, Tatjana Kleine
ABSTRACT
Singlet oxygen (1O2), the major reactive oxygen species (ROS) produced in chloroplasts, has been demonstrated recently to be a highly versatile signal that induces various stress responses. In the fluorescent (flu) mutant, its release causes seedling lethality and inhibits mature plant growth. However, these drastic phenotypes are suppressed when EXECUTER1 (EX1) is absent in the flu ex1 double mutant. We identified SAFEGUARD1 (SAFE1) in a screen of ethyl methanesulfonate (EMS) mutagenized flu ex1 plants for suppressor mutants with a flu-like phenotype. In flu ex1 safe1, all 1O2-induced responses, including transcriptional rewiring of nuclear gene expression, return to levels, such as, or even higher than, those in flu Without SAFE1, grana margins (GMs) of chloroplast thylakoids (Thys) are specifically damaged upon 1O2 generation and associate with plastoglobules (PGs). SAFE1 is localized in the chloroplast stroma, and release of 1O2 induces SAFE1 degradation via chloroplast-originated vesicles. Our paper demonstrates that flu-produced 1O2 triggers an EX1-independent signaling pathway and proves that SAFE1 suppresses this signaling pathway by protecting GMs.
Sigma-Aldrich
Anti-Green Fluorescent Protein (GFP) antibody, Mouse monoclonal, clone GSN149, purified from hybridoma cell culture
Millipore
Immobilon®-P PVDF Membrane, 1 roll, 27 cm x 3.75 m, 0.45 µm pore size, Hydrophobic PVDF Transfer Membrane for western blotting.