Merck
  • Dose-Dependent Alterations to In Vitro Human Microbiota Composition and Butyrate Inhibition by a Supercritical Carbon Dioxide Hops Extract.
Dose-Dependent Alterations to In Vitro Human Microbiota Composition and Butyrate Inhibition by a Supercritical Carbon Dioxide Hops Extract.
Biomolecules (2019-08-24)
Paul A Blatchford, Shanthi G Parkar, Wendy Hopkins, John R Ingram, Kevin H Sutton
ABSTRACT
Hop cones (Humulus lupulus L.) have been used throughout history as an additive in beer brewing and as herbal supplements with medicinal and culinary properties. The objective of this study was to ascertain the effect of a range of concentrations of a supercritical CO2 extract of hops on the composition and metabolism of human gut bacterial communities using in vitro batch culture systems. Fermentations were conducted over 24 h using a mixed human fecal inoculum. Microbial metabolism was assessed by measuring organic acid production and microbial community alterations were determined by 16S rRNA gene sequencing. Butyrate, an important short chain fatty acid in maintaining colonic well-being, decreased at elevated concentrations of hops, which may partly be accounted for by the concomitant reduction of Eubacterium and Coprococcus, known butyrate-producing genera, and also the inhibition of Bifidobacterium, a beneficial organism that has a butyrogenic effect through metabolic cross-feeding with intestinal commensals. The hops compounds also caused dose-dependent increases in the potentially pathogenic Enterobacteriaceae and potentially beneficial Akkermansia. Thus, hops compounds had a significant impact on the structure of the bacterial consortium, which warrants further study including human clinical trials.