Merck
Passively Addressable Ultra-Low Volume Sweat Chloride Sensor.
Sensors (Basel, Switzerland) (2019-10-28)
Antra Ganguly, Shalini Prasad
ABSTRACT
This work demonstrates a novel electrochemical biosensor for the detection of chloride ion levels in ultra-low volumes (1-3 microliters) of passively expressed human sweat. We present here a hydration monitor that the pediatric, geriatric, and other immune-compromised or physically inactive/sedentary population cohort can utilize, for whom the current methods of chloride quantification of active stimulation of sweat glands through iontophoresis or treadmill runs are unsuitable. In this work, non-faradaic electroanalysis using gold microelectrodes deposited on a flexible nanoporous substrate, for high nanoscale surface area to volume enhancement, was leveraged to operate in ultra-low sweat volumes of <3 µL eluted at natural rates. The specific chloride ionophore-based affinity of chloride ions resulted in the modulation of charge transfer within the electrical double layer at the electrode-sweat buffer interface, which was transduced using electrochemical impedance spectroscopy (EIS) and chronoamperometry (CA). Linear calibration dose responses with R-squared values of 0.9746 and 0.9403 for EIS and CA respectively were obtained for a dynamic range of 10-100 mM. The surface charge and the binding chemistry of the capture probe were studied using zeta potential studies and UV-Vis. The dynamic sweat chloride-tracking capability of the sensor was evaluated for a duration of 180 min. Studies were conducted to probe the efficacy of the developed sensor for passive ultra-low sweat chloride assessment on human subjects (n = 3).
Product Description
Supelco
Chloride ionophore IV, Selectophore, for ion-selective electrodes